Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraComp eBeads™ Plus Compensation Beads

Catalog No. 01333342
Encompass_Preferred
Change view
Click to view available options
No. of Tests:
25
100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
01-333-342 100
01-333-341 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-333-342 Supplier Invitrogen™ Supplier No. 01333342
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

UltraComp eBeads Plus Compensation Beads are the second generation of UltraComp beads for use in flow cytometry.

  • Binds the spherical particles for use as a single-color compensation controls.
  • Binds with mouse, rat and hamster antibodies, but also bind with rabbit and recombinant human antibodies.
  • Designed for use in compensation with all fluorochromes excited by ultraviolet (355 nm) violet (405 nm), blue (488 nm), green (532 nm), yellow-green (561 nm), and red (633-640 nm) lasers.
  • Compensate in a manner similar to cells when used with Brilliant Violet 785/786-, Brilliant Violet 711-, Super Bright 780- and Super Bright 702-conjugated antibodies.
  • Multispecies compatibility—bind with mouse, rat, hamster, rabbit and recombinant human antibodies
  • Improved fluorophore compatibility
  • Each drop of beads contains two populations: a positive population that bind with any mouse, rat, hamster, rabbit or recombinant human antibody and a negative population that will not bind with antibody.
  • When a fluorochrome-conjugated antibody is added to the beads, both positive and negative populations are observed.
  • This bimodal distribution can be used for single-color compensation controls to accurately setup instrumentation for multicolor flow cytometry experiments.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 tests
Product Type Compensation Beads
Diameter (Metric) 5 μm
Excitation/Emission UV-633/635 nm
For Use With (Application) Mouse, rat, hamster, human, rabbit antibodies
No. of Tests 100
Concentration 5 x 106 beads/mL
What is the size of UltraComp eBeads Plus Compensation Beads?

The bead size is 5 µm.

RUO use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.