Learn More
Santa Cruz Biotechnology ELISA Plate, High Binding, 96 well, Flat Bottom, 40/cs
Supplier:  Santa Cruz Biotechnology SC204463
Encompass
96 well ELISA plates are manufactured from clear, virgin polystyrene and selected for optical clarity and consistency. ELISA plates are formatted with MICROLON 600 for high binding assays. Plates come with flat bottom wells. 96 well ELISA plates come with a chimney-well design option that reduces the risk of cross-contamination and facilitates sealing by cap mat, thermal sealer, or adhesive sealer.
Catalog No. NC9007188