Learn More
Santa Cruz Biotechnology ULTRACRUZ LAB TAPE VIOLET 4PK
Catalog No. NC0070241
Encompass
Violet colored labeling tape. 0.75 in. (19 mm) wide. Withstands temperatures from -23 to +121 Deg.C. Will stick to most surfaces . Write on it with pen, pencil or permanent marker. Sold as 60 yd. (55 m) rolls; 4 per pack. Waterproof, oil and acid resistant.
Catalog No. NC0070241 Supplier Santa Cruz Biotechnology Supplier No. SC224492
Violet colored labeling tape. 0.75 in. (19 mm) wide. Withstands temperatures from -23 to +121 Deg.C. Will stick to most surfaces . Write on it with pen, pencil or permanent marker. Sold as 60 yd. (55 m) rolls; 4 per pack. Waterproof, oil and acid resistant.