Learn More
Santa Cruz Biotechnology UltraCruz Lab Tape, Yellow, 4/pk
Catalog No. NC0070234
Encompass
Lab tape is strong, durable, waterproof, oil and acid resistant which is suitable for various labeling applications. Lab tape will stick to most surfaces including glass, plastic and metal. Lab tape can withstand temperatures from -23 to +121°C. 3/4"""" (19 mm) wide roll. Tape can be written on with pen, pencil or permanent marker.
Catalog No. NC0070234 Supplier Santa Cruz Biotechnology Supplier No. SC224491
Lab tape is strong, durable, waterproof, oil and acid resistant which is suitable for various labeling applications. Lab tape will stick to most surfaces including glass, plastic and metal. Lab tape can withstand temperatures from -23 to +121°C. 3/4"""" (19 mm) wide roll. Tape can be written on with pen, pencil or permanent marker.