Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife 3.0 mm, 35° Knife angle
SDP
Catalog No. 501898812
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 35°
Catalog No. 50-189-8812 Supplier Electron Microscopy Sciences Supplier No. 30UL
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 35°