Learn More
Ted Pella Inc Ultrafine Tungsten Manipulator Probe
Supplier: Ted Pella Inc 13570
The Ultrafine Tungsten Manipulator Probe consists of a fine tungsten needle with 0.6um radius in a quality precision pin vise. The ultrafine point is ideally suitable for probing and manipulating very small particles or feature under a microscope. The tungsten needle has a diameter of 0.52mm (0.020"), a length of 25mm (1") and a sharp tip radius of 0.6µm. The precision pin vise has knurled brass handle for a firm grip and a precision carbon steel collet for holding the tungsten probe. 80mm (3-1/8") long. The maximum probe diameter for the pin vise is 0.70mm (0.028"). The sharp tungsten needle can also be used in other needle holders or for custom made probing systems.