Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ UltraFIT™ Nylon Laboratory/Cleanroom Glove Liners

Catalog No. p-7230586 Shop All Epredia Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
14-379-36 Medium
14-379-45 Large
19-130-4560 X-Large
14-379-35 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 14-379-36 Supplier Epredia™ Supplier No. 5100212001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pre-shrunk nylon glove liners protect extra-sensitive hands from irritation and occlusion of the skin associated with frequent gloving.

  • Pre-shrunk nylon is unaffected by repeated laundering
  • Seamless fingertips ensure optimum dexterity
  • Protects extra sensitive hands from irritation and occlusion of the skin
  • Use to provide an extra layer of warmth when working in cold areas
  • Color coded hems for easy size identification

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Medium
For Use With (Application) Preventing Irritation and Skin Occlusion
Glove Cuff Style Knit
Series UltraFIT™
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.