Learn More
Innovative Med Tech UltraFlo-A Luer Access Device
Catalog No. 22757230
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
22-757-230 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 22-757-230 Supplier Innovative Med Tech Supplier No. 902
Add to Cart
Add to Cart
This item is not returnable. View return policy
The UltraFlo-A Luer Access Device is a pre-assembled blood tube holder with a multisample male luer adapter.
  • A sterile, pre-assembled blood collection tube holder with integrated multi-sample luer adapter
  • Compatible with female luer connections or IV ports designed for luer access
  • Blue color-coded connection provides easy differentiation from other holder-based products
Specifications
Material Polypropylene
Autoclavable Not Autoclavable
Color Blue
Connectors Male luer
Product Type Blood Collection Access Device
Disposable Single-use
Latex Free Yes
Sterility Sterile