Learn More
Innovative Med Tech UltraFlo-C Capillary Collector K2 EDTA
Catalog No. 21403425
Change view
Click to view available options
Capacity (Metric):
100 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
21-403-425 200 μL
21-403-424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-425 Supplier Innovative Med Tech Supplier No. 682005
The ideal combination of controlled blood collection and sample container to avoid sample loss or contamination.
  • The ideal combination of controlled blood collection and sample container to avoid sample
  • Exact measurement and stabilization of the required amount of blood via integrated end-to-end plastic capillary coated with anti-coagulant.
  • Reduction of the infection risk thanks to unbreakable plastic capillaries.
  • Clear marking of expiry date and production batch on each sample container
Specifications
Color Purple
Product Type Capillary Tube
Additive K2 EDTA
Material Polypropylene
Sterility Non-sterile
Closure Type Snap-lock
Capacity (Metric) 200 μL