Learn More
Innovative Med Tech UltraFlo-R Push Button Blood Collection and Infusion Set
Catalog No. p-200006999
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Color:
Blue
Green
Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Color
22-757-235 21 G Green
22-757-236 23 G Blue
22-757-237 25 G Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-757-235 Supplier Innovative Med Tech Supplier No. 365
Add to Cart
Add to Cart
This item is not returnable. View return policy
UltraFlo-R Push Button Blood Collection and Infusion Set allows for safety with the push of a button. Single-handed activation retracts the needle instantly from the vein into the needle hub. Ultraflo-R needle increases flow rate.
  • Sterile, single use, butterfly, multi-sample, blood collection and infusion set
  • Single-handed activation retracts the needle instantly from the vein into the needle hub
  • Minimizes sharp exposure
  • UltraFlo needle cannula increases flow rate
  • Improve patient comfort: Use 23G instead of 21G without compromising flow rate
  • Approved for short-term infusion (<2 hrs)
  • Unique bevel design requires lower penetration force*, providing for a more comfortable patient experience (*Compared to leading-brand standard cannula)
Specifications
Color Green
Material PVC
Needle Gauge 21 G
Sterility Sterile
Latex Free Yes