Learn More
Innovative Med Tech UltraFlo-S Push Button Blood Collection/Infusion Set, with Holder attached
Catalog No. 22757241
Change view
Click to view available options
Needle Gauge:
25 G
Color:
Orange
Catalog No. Needle Gauge Color
22-757-241 25 G Orange
1 options
This item is not returnable. View return policy
Catalog No. 22-757-241 Supplier Innovative Med Tech Supplier No. 656
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ultraflo™ needle increases flow rate and improves patient comfort.
  • One or two-handed safety shield blood collection/infusion set. 
Specifications
Color Orange
Material PVC
Needle Gauge 25 G
Sterility Sterile
Latex Free Yes
Length (English) Needle 0.75 in.
Length (English) Tube 12 in.