Learn More
Innovative Med Tech UltraFlo-T Blood Transfer Device
Description
- A sterile, pre-assembled and easy-to-use device designed with safety in mind
- Used for needleless specimen transfer from a syringe to an evacuated tube or blood culture bottle
- Pink color-coded connection provides easy differentiation from other holder-based products
Specifications
Specifications
| Material | Polypropylene |
| Autoclavable | Not Autoclavable |
| Color | Pink |
| Connectors | Female luer |
| Product Type | Blood Transfer Device |
| Disposable | Single-use |
| Latex Free | Yes |
| Sterility | Sterile |