Learn More
Research Products International Corp UltraFlux 96-Well Plate, Semi-Skirted Bottom, with Raised Rim, 10 per Package
SDP
Catalog No. 501978225
Encompass
These plates are designed for use with all popular brands of thermal cyclers and sequence detection systems. Plates are semi-skirted with "V" shaped wells and a notched corner at A12 position. This plate has indented sides with a raised top rim around the plates. Plates can be sealed with film, caps or mats.
Catalog No. 50-197-8225 Supplier Research Products International Corp Supplier No. 248425
These plates are designed for use with all popular brands of thermal cyclers and sequence detection systems. Plates are semi-skirted with "V" shaped wells and a notched corner at A12 position. This plate has indented sides with a raised top rim around the plates. Plates can be sealed with film, caps or mats.