Learn More
Research Products International Corp UltraFlux 96 Well Plate, Straight Sides, 10/pk
SDP
Catalog No. 501978224
Encompass
These plates are designed for use with all popular brands of thermal cyclers and sequence detection systems. Plates are semi-skirted with "V" shaped wells and a notched corner at A12 position. This UltraFlux plate is rimless with straight sides for compatibility with newer sequencing equipment. Plates can be sealed with film, caps or mats.
Catalog No. 50-197-8224 Supplier Research Products International Corp Supplier No. 248450
These plates are designed for use with all popular brands of thermal cyclers and sequence detection systems. Plates are semi-skirted with "V" shaped wells and a notched corner at A12 position. This UltraFlux plate is rimless with straight sides for compatibility with newer sequencing equipment. Plates can be sealed with film, caps or mats.