Learn More
Research Products International Corp UltraFlux 96 Well PCR Plate with Skirted Bottom, Natural, 10 per Package
SDP
Catalog No. 50212625
Encompass
UltraFlux 96-well 0.2 ml PCR Plate simplifies sample processing and saves time. Minimizes errors and improves efficiency compared to individual tube strips. UltraFlux PCR plates feature a skirted bottom for bar coding. Slightly textured surface allows easy removal from the cycler block. Ideal for use in high throughput robotic systems or for individual plate runs. Fits all popular brands of thermal cyclers. Plates can be sealed with sealing film or 0.2 ml cap strips. Manufactured from polypropylene. PCR plates are non-sterile.
Catalog No. 50-212-625 Supplier Research Products International Corp Supplier No. 248861
UltraFlux 96-well 0.2 ml PCR Plate simplifies sample processing and saves time. Minimizes errors and improves efficiency compared to individual tube strips. UltraFlux PCR plates feature a skirted bottom for bar coding. Slightly textured surface allows easy removal from the cycler block. Ideal for use in high throughput robotic systems or for individual plate runs. Fits all popular brands of thermal cyclers. Plates can be sealed with sealing film or 0.2 ml cap strips. Manufactured from polypropylene. PCR plates are non-sterile.