Learn More
Research Products International Corp UltraFlux Low Profile PCR Tubes, White, 960/cs
SDP
Catalog No. 501977906
Encompass
UltraFlux Low Profile PCR Tubes are supplied on a convenient 8 tube strip with flat caps attached at an offset angle. Low profile tubes feature precision molded thin walls that provide maximum heat transfer and a shorter length that reduces unnecessary head space. Normal working capacity of each tube is 100ul and a total capacity of 200ul. Tube strips are free from DNase, RNase, DNA, endotoxins and PCR inhibitors. Tubes available in white for optimum reflectance of fluorescent signal in QPCR techniques. Caps have an optically clear flat surface.
Catalog No. 50-197-7906 Supplier Research Products International Corp Supplier No. 249970
UltraFlux Low Profile PCR Tubes are supplied on a convenient 8 tube strip with flat caps attached at an offset angle. Low profile tubes feature precision molded thin walls that provide maximum heat transfer and a shorter length that reduces unnecessary head space. Normal working capacity of each tube is 100ul and a total capacity of 200ul. Tube strips are free from DNase, RNase, DNA, endotoxins and PCR inhibitors. Tubes available in white for optimum reflectance of fluorescent signal in QPCR techniques. Caps have an optically clear flat surface.