Learn More
Research Products International Corp UltraFlux Rigid 0.2mL PCR Tube Strips, White, 960pk
SDP
Catalog No. 501977790
Encompass
UltraFlux Rigid PCR Tube Strips are easier to work with than strips designed to snap apart. These eight thin wall tubes are joined together by a strong double link that reduces flexibility and provides more convenient handling. White color tubes maximize reflectance and enhance the signal.
Catalog No. 50-197-7790 Supplier Research Products International Corp Supplier No. 249951
UltraFlux Rigid PCR Tube Strips are easier to work with than strips designed to snap apart. These eight thin wall tubes are joined together by a strong double link that reduces flexibility and provides more convenient handling. White color tubes maximize reflectance and enhance the signal.