Learn More
Research Products International Corp UltraFlux EZ Hinge PCR Tube Strips, Clear, 125/pk
SDP
Catalog No. 501977791
Encompass
UltraFlux PCR EZ Tube Strips feature a unique hinge design that allows fast and convenient opening and closing of tubes. Optically clear caps for real-time PCR(qPCR).
Size: 0.2 ml
Catalog No. 50-197-7791 Supplier Research Products International Corp Supplier No. 249955
Add to Cart
Add to Cart
UltraFlux PCR EZ Tube Strips feature a unique hinge design that allows fast and convenient opening and closing of tubes. Optically clear caps for real-time PCR(qPCR).
Size: 0.2 ml