Learn More
Electron Microscopy Sciences Gold perforated support foil with pre-defined hole size, shape and arrangement. It has advantages in cryo electron microscopy reducing the movement of frozen specimens leading to better 3D reconstructions.
SDP
Catalog No. NC2257089
Encompass_Preferred
Newly developed ultrastable gold supports for electron cryomicroscopy will reduce the movement of frozen specimens during imaging. This improves image contrast and quality leading to better 3D reconstructions with less data. During imaging at cryo-temperatures, traditional carbon supports move, particularly at the beginning of irradiation. This movement blurs images and makes it difficult to determine the structures of small and challenging molecules.UltrAuFoilswere designed at MRC's Laboratory of Molecular Biology by Dr. Christopher J. Russo and Dr. Lori A. Passmore.
This item is not returnable. View return policy
Catalog No. NC2257089 Supplier Electron Microscopy Sciences Supplier No. Q350AR13A
Newly developed ultrastable gold supports for electron cryomicroscopy will reduce the movement of frozen specimens during imaging. This improves image contrast and quality leading to better 3D reconstructions with less data. During imaging at cryo-temperatures, traditional carbon supports move, particularly at the beginning of irradiation. This movement blurs images and makes it difficult to determine the structures of small and challenging molecules.UltrAuFoilswere designed at MRC's Laboratory of Molecular Biology by Dr. Christopher J. Russo and Dr. Lori A. Passmore.