Learn More
Perkin Elmer US LLC Ultra-High Purity 98mL Disposable Helium Gas Cylinder: Purity 99.5% (12/pack)
Catalog No. 502113806
Encompass_Preferred
Disposable helium gas cylinder is a 98 mL stainless steel cylinder at 2500 psi. Helium cylinders are used for both the T-9 GC/MS and SPS-3 during portable operation.
Catalog No. 50-211-3806 Supplier Perkin Elmer US LLC Supplier No. NTSSG070037
Disposable helium gas cylinder is a 98 mL stainless steel cylinder at 2500 psi. Helium cylinders are used for both the T-9 GC/MS and SPS-3 during portable operation.