Learn More
Perkin Elmer US LLC Ultra-High Purity 98mL Disposable Helium Gas Cylinder: Purity 99.95% (12/pack)
Catalog No. 502113791
Encompass_Preferred
Disposable helium gas cylinder is a 98 mL stainless steel cylinder at 2500 psi. Helium cylinders are used for both the T-9 GC/MS and SPS-3 during portable operation.
Catalog No. 50-211-3791 Supplier Perkin Elmer US LLC Supplier No. NTSST090037
Add to Cart
Add to Cart
Request a Quote
Disposable helium gas cylinder is a 98 mL stainless steel cylinder at 2500 psi. Helium cylinders are used for both the T-9 GC/MS and SPS-3 during portable operation.