Learn More
Waters Corp Waters Column, 1000 Å, 12 μm, 7.8 mm × 300 mm
Catalog No. 50785003 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Ultrahydrogel Columns are aqueous-compatible gel columns for analytical and preparative separations of water-soluble polymers, including oligomers, polysaccharides, nucleic acids, proteins, and peptides.
  • Sophisticated aqueous-compatible gel column for analytical and preparative separations.
  • Characterizes oligomers and biological substances.
  • Enables characterization of cationic, anionic, and amphoteric polymers.
  • Offers high resolution for aqueous SEC columns.
  • Wide working pH range of 2 to 12.
  • Compatible with high concentrations of organic solvents.
  • Provides high resolution, low adsorption, and wide molecular weight separations within 10°C to 80°C.
  • Manufactured in ISO 9001 certified facility.
Catalog No. 50-785-003 Supplier Waters Corp Supplier No. WAT011535
Ultrahydrogel Columns are aqueous-compatible gel columns for analytical and preparative separations of water-soluble polymers, including oligomers, polysaccharides, nucleic acids, proteins, and peptides.
  • Sophisticated aqueous-compatible gel column for analytical and preparative separations.
  • Characterizes oligomers and biological substances.
  • Enables characterization of cationic, anionic, and amphoteric polymers.
  • Offers high resolution for aqueous SEC columns.
  • Wide working pH range of 2 to 12.
  • Compatible with high concentrations of organic solvents.
  • Provides high resolution, low adsorption, and wide molecular weight separations within 10°C to 80°C.
  • Manufactured in ISO 9001 certified facility.