Learn More
Waters Corp Ultrahydrogel Column, 120Å, 6 µm, 7.8 mm X 300 mm, 100 - 5K, 1/pk
Supplier:  Waters Corp WAT011520
Encompass_Preferred
Ideal for the analysis of aqueous-soluble samples, such as; oligomers; oligosaccharides; polysaccharides; and cationic, anionic and amphoteric polymers.
Catalog No. 50-785-000