Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Ultrahydrogel Column, 2000Å, 12 μm, 7.8 mm x 300 mm

Catalog No. 50785004 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Ultrahydrogel columns are aqueous-compatible gel columns designed for analytical and preparative separations of water-soluble polymers. They are suitable for characterizing oligomers, oligosaccharides, and various biological components including polysaccharides, nucleic acids, proteins, and peptides.

  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Temperature limits: 10°C to 80°C
  • Molecular weight range: 50000 to 7000000
  • Particle shape: Spherical
  • Particle size: 12 μm
  • Pore size: 2000 Å
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • System: HPLC
  • Technique: SEC
  • High resolution
  • Broad working pH range
  • Flexible mobile phase options with fewer non-size exclusion effects
  • Compatible with high concentrations of organic solvents (up to 50%)

Catalog No. 50-785-004 Supplier Waters Corp Supplier No. WAT011540
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Ultrahydrogel columns are aqueous-compatible gel columns designed for analytical and preparative separations of water-soluble polymers. They are suitable for characterizing oligomers, oligosaccharides, and various biological components including polysaccharides, nucleic acids, proteins, and peptides.

  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Temperature limits: 10°C to 80°C
  • Molecular weight range: 50000 to 7000000
  • Particle shape: Spherical
  • Particle size: 12 μm
  • Pore size: 2000 Å
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • System: HPLC
  • Technique: SEC
  • High resolution
  • Broad working pH range
  • Flexible mobile phase options with fewer non-size exclusion effects
  • Compatible with high concentrations of organic solvents (up to 50%)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.