Learn More
Waters Corp Waters Ultrahydrogel DNA Column, 10 μm, 7.8 mm x 300 mm
Catalog No. 50785007 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Ultrahydrogel Columns contain a sophisticated aqueous-compatible gel for analytical and preparative separations of water-soluble polymers. The gel is a cross-linked hydroxylated polymer with some residual carboxyl functionality. These columns are used to characterize oligomers and biological substances such as polysaccharides, nucleic acids, proteins, and peptides.
  • High-resolution separations.
  • Wide pH range (2 to 12 pH).
  • Compatibility with high concentrations of organic solvents.
  • Higher mobile phase flexibility.
  • Lower incidence of non-size-exclusion effects.
  • Manufactured under strict controls to ensure adherence to high industry standards.
Catalog No. 50-785-007 Supplier Waters Corp Supplier No. WAT011560
Waters Ultrahydrogel Columns contain a sophisticated aqueous-compatible gel for analytical and preparative separations of water-soluble polymers. The gel is a cross-linked hydroxylated polymer with some residual carboxyl functionality. These columns are used to characterize oligomers and biological substances such as polysaccharides, nucleic acids, proteins, and peptides.
  • High-resolution separations.
  • Wide pH range (2 to 12 pH).
  • Compatibility with high concentrations of organic solvents.
  • Higher mobile phase flexibility.
  • Lower incidence of non-size-exclusion effects.
  • Manufactured under strict controls to ensure adherence to high industry standards.