Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, 120 Å, 6 μm, 7.8 mm X 300 mm

Catalog No. 50785006 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Waters Ultrahydrogel Columns provide aqueous-compatible gel separation for water-soluble polymers. They characterize oligomers, polysaccharides, and peptides, offering high resolution, low adsorption, and mobile phase flexibility, even with high organic solvent concentrations.

  • Chemistry: SEC
  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Temperature limits: 10°C to 80°C
  • Molecular weight range: 100 to 5000
  • Particle shape: Spherical
  • Particle size: 6 μm
  • Endfitting type: Waters
  • Pore size: 120 Å
  • Format: Column
  • Packing solvent: Water
  • System: HPLC
  • Technique: SEC
  • USP classification: L25
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Application: Gel filtration
  • Units per package: 1 pk

Catalog No. 50-785-006 Supplier Waters Corp Supplier No. WAT011550
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Waters Ultrahydrogel Columns provide aqueous-compatible gel separation for water-soluble polymers. They characterize oligomers, polysaccharides, and peptides, offering high resolution, low adsorption, and mobile phase flexibility, even with high organic solvent concentrations.

  • Chemistry: SEC
  • Separation mode: Gel filtration (aqueous)
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Temperature limits: 10°C to 80°C
  • Molecular weight range: 100 to 5000
  • Particle shape: Spherical
  • Particle size: 6 μm
  • Endfitting type: Waters
  • Pore size: 120 Å
  • Format: Column
  • Packing solvent: Water
  • System: HPLC
  • Technique: SEC
  • USP classification: L25
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Application: Gel filtration
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.