Learn More
Waters Corp Waters Corporation Guard Column, 6 μm Particle, 6 mm ID, 40 mm Length
Catalog No. 50785009 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Ultrahydrogel Columns contain a sophisticated aqueous-compatible gel for analytical and preparative separations of water-soluble polymers. The gel is a cross-linked hydroxylated polymer with some residual carboxyl functionality. Ultrahydrogel DP also contains some residual amine functionality. Compounds normally characterized include oligomers and biological substances such as polysaccharides, nucleic acids, proteins, and peptides.
  • Designed to extend the lifespan of the analytical column.
  • Constructed using the same high-performance packing materials as the analytical column.
  • Functions by preventing suspended particles and chemical contaminants from entering the analytical column.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch of packing material is chromatographically examined to guarantee the highest quality and ensures good, repeatable performance.
Catalog No. 50-785-009 Supplier Waters Corp Supplier No. WAT011570
Ultrahydrogel Columns contain a sophisticated aqueous-compatible gel for analytical and preparative separations of water-soluble polymers. The gel is a cross-linked hydroxylated polymer with some residual carboxyl functionality. Ultrahydrogel DP also contains some residual amine functionality. Compounds normally characterized include oligomers and biological substances such as polysaccharides, nucleic acids, proteins, and peptides.
  • Designed to extend the lifespan of the analytical column.
  • Constructed using the same high-performance packing materials as the analytical column.
  • Functions by preventing suspended particles and chemical contaminants from entering the analytical column.
  • Produced in a cGMP, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch of packing material is chromatographically examined to guarantee the highest quality and ensures good, repeatable performance.