Learn More
Waters Corp Waters Ultrahydrogel Guard Column, 125Å Pore, 6 μm Particle
Supplier:  Waters Corp WAT011565
Encompass_Preferred
This Ultrahydrogel Guard Column provides an aqueous-compatible gel for analytical and preparative separations of water-soluble polymers, including oligomers, biological substances, polysaccharides, nucleic acids, proteins, and peptides. Installed between the injector and analytical column, it removes impurities and suspended solids, extending column life and reducing lab costs.
  • Extends analytical column lifetime.
  • Utilizes high-performance packing materials identical to the analytical column.
  • Filters out chemical impurities and suspended solids.
  • Produced in a cGMP, ISO 9001 certified plant with ultra-pure reagents.
  • Batch tested chromatographically for quality and reproducible performance.
  • Lowers lab equipment costs by reducing column replacement frequency.
Catalog No. 50-785-008