Learn More
Sigma Aldrich Ultral-thin Carbon Film Supported Copper Gilder Finder F1 Grids 200 Mesh 50/box
Catalog No. 503126053 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 Pk.
Catalog No. Quantity
50-312-6053 1 Pk.
1 options
Catalog No. 50-312-6053 Supplier Sigma Aldrich Supplier No. TEMCF200F1CUUL
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Ultral-thin Carbon Film Supported Copper Gilder Finder F1 Grids 200 Mesh 50/box
Specifications
Quantity 1 Pk.