Learn More
Myron L Company Myron L Ultrameter II Model 4P, Accurate Conductivity Measurement, Waterproof Design, 12.4 oz
SDP
Catalog No. NC3432941
Encompass
The Myron L Company Ultrameter II Model 4P is a versatile handheld instrument for accurately measuring conductivity, TDS, resistivity, and temperature, making it ideal for scientific and industrial applications. Its waterproof design, advanced four-electrode technology, and over 100 hours of battery life enhance reliability and usability in the field. With data logging capabilities and an intuitive display, it stands out among similar products by providing high accuracy and ease of use.
Catalog No. NC3432941 Supplier Myron L Company Supplier No. 4PIIFCE
The Myron L Company Ultrameter II Model 4P is a versatile handheld instrument for accurately measuring conductivity, TDS, resistivity, and temperature, making it ideal for scientific and industrial applications. Its waterproof design, advanced four-electrode technology, and over 100 hours of battery life enhance reliability and usability in the field. With data logging capabilities and an intuitive display, it stands out among similar products by providing high accuracy and ease of use.