Learn More
Ted Pella Inc ULTRAMICRO-NEEDLE,30DEG,0.50MM
SDP
Catalog No. NC1807925
Encompass
Ultra Micro-Needle, tungsten, 30° tip radius 5 µm, 0.50mm (0.020")
Catalog No. NC1807925 Supplier Ted Pella Inc Supplier No. 13626B
Ultra Micro-Needle, tungsten, 30° tip radius 5 µm, 0.50mm (0.020")