Learn More
Ted Pella Inc ULTRAMICRO-NEEDLE,TGSTN,0.25MM
SDP
Catalog No. NC1807922
Encompass
Ultra Micro-Needle, tungsten, straight, tip radius 5 µm, 0.25mm (0.010")
Catalog No. NC1807922 Supplier Ted Pella Inc Supplier No. 13625A
Ultra Micro-Needle, tungsten, straight, tip radius 5 µm, 0.25mm (0.010")