Learn More
World Precision Instrument UMP3 UltraMicroPump -UMP3-3
SDP
Catalog No. NC2455781
Encompass
With its digital controller, UltraMicroPump III can repeatably inject as little as 25nL. Syringes may be filled externally and then inserted into the pump or front-filled while mounted on the pump. 
Catalog No. NC2455781 Supplier World Precision Instrument Supplier No. UMP33
Add to Cart
Add to Cart
Request a Quote
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
With its digital controller, UltraMicroPump III can repeatably inject as little as 25nL. Syringes may be filled externally and then inserted into the pump or front-filled while mounted on the pump. 