Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Ultramicrotomy Grade, 8 mm thick, 25 x 400 mm
SDP
Catalog No. 50930671
Encompass_Preferred
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 8 mm
Size: 25 x 400 mm
Catalog No. 50-930-671 Supplier Electron Microscopy Sciences Supplier No. 71013
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 8 mm
Size: 25 x 400 mm