Learn More
Foamtec UltraMOP™ II Mop Heads
Catalog No. 19130420 Shop All Foamtec International Products
Change view
Click to view available options
Product Type:
Mop Head
Sterility:
Sterile
Catalog No. Product Type Sterility
19-130-420 Mop Head Sterile
1 options
Catalog No. 19-130-420 Supplier Foamtec Supplier No. FS505B1ST
Add to Cart
Add to Cart
Designed for wet mopping procedures in sterile and controlled environments
Constructed from a fine-pore urethane foam to enhance application of disinfectants to clean room surfaces. Available in laminated versions to allow tear-free cleaning and disinfecting of areas that have rough or sharp surfaces.
  • Proprietary polyurethane foam compatible with disinfectants used for maintaining sterile environments
  • Laminated bump-proof designs enable tear-free cleaning and disinfecting of areas that have rough or sharp surfaces
  • Micro-fiber versions are designed to remove and absorb stubborn contamination such as hardened biofilms and disinfectant residue, perfect for drying floors
  • Individually packaged to maintain cleanliness
  • Fine, uniform pore size and proprietary, patent-pending, lamination techniques ensure excellent laydown and pickup properties
  • Available with lot Certification of Conformance for ease of audit and validation procedures
Specifications
Product Type Mop Head
Length (English) 16 in.
Sterility Sterile
Includes Polyester Lamination
For Use With (Application) Designed for wet mopping procedures in sterile and controlled environments
Material Urethane Foam