Learn More
Electron Microscopy Sciences ULTRAPOL Basic Manual Polisher
SDP
Catalog No. NC1634594
Encompass
Manual polishing tool for delayering and/or crosssectioning results on a very tight budget. Works with all 8" (200 mm) surfaces. Features a small footprint, speed control and high torque motor, and coolant speed control. Configured for 100-220/240V operation. 0-500 RPM. Includes 8 inch (200 mm) polishing system with speed control and solenoid-controlled coolant system.
Catalog No. NC1634594 Supplier Electron Microscopy Sciences Supplier No. 1209.1
Manual polishing tool for delayering and/or crosssectioning results on a very tight budget. Works with all 8" (200 mm) surfaces. Features a small footprint, speed control and high torque motor, and coolant speed control. Configured for 100-220/240V operation. 0-500 RPM. Includes 8 inch (200 mm) polishing system with speed control and solenoid-controlled coolant system.