Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UltraPure™ Cesium Chloride, Optical Grade

Catalog No. 15507023
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-507-023 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-507-023 Supplier Invitrogen™ Supplier No. 15507023
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients.

UltraPureTM Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPureTM Cesium Chloride, Optical Grade can be used to prepare plasmids or to isolate phage, virus particles, or cellular RNA.

UltraPure™ Cesium Chloride, Optical Grade is specifically prepared for use in density gradients. The highest grade commercially available, UltraPure™ Cesium Chloride, Optical Grade can be used to prepare plasmids or to isolate phage, virus particles, or cellular RNA.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cesium Chloride
Content And Storage Store at room temperature.
Product Line UltraPure
Quantity 1 kg
Shipping Condition Room Temperature
Form Powder

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.