Learn More
Invivogen Ultrapure flagellin from B . Subtilis, 50 ug
SDP
Catalog No. NC0611139
Encompass
Flagellin, the principal component of the flagella present on many Gram-negative and Gram-positive bacteria, is a proinflammatory molecule recognized by distinct types of pattern recognition receptors (PRRs); the surface localized Toll-like receptor (TLR5) and the cytosolic NOD-like receptors (NLRs), NLRC4 and NAIP5.
Catalog No. NC0611139 Supplier Invivogen Supplier No. TLRLPBSFLA
Flagellin, the principal component of the flagella present on many Gram-negative and Gram-positive bacteria, is a proinflammatory molecule recognized by distinct types of pattern recognition receptors (PRRs); the surface localized Toll-like receptor (TLR5) and the cytosolic NOD-like receptors (NLRs), NLRC4 and NAIP5.