Learn More
Eurofins Discoverx Ultrapure IBMX, 55.55 mg. (92-0007)
Supplier:  Eurofins Discoverx 920007
Encompass
IBMX, also referred to as 3-isobutyl-1-methylxanthine, is a competitive phosphodiesterase inhibitor, thereby slowing degradation of intracellular cAMP; activates PKA; inhibits TNF-alpha; synthesizes leukotrienes, and reduces a number of inflammation and innate immune mechanisms. IBMX also is known among the class of compounds to act as a nonselective adenosine receptor antagonist. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9545