Learn More
Invivogen ULTRAPURE LIPOPOLYSACCHARIDE F
SDP
Catalog No. NC0065722
Encompass
Lipopolysaccharide from Escherichia coli K12 Ultra-pure LPS-EK was extracted by successive enzymatic hydrolysis steps and purified by the phenol-TEA-DOC extraction protocol. Ultrapure LPS-EK only activates the TLR4 pathway.
Catalog No. NC0065722 Supplier Invivogen Supplier No. TLRLPEKLPS
Lipopolysaccharide from Escherichia coli K12 Ultra-pure LPS-EK was extracted by successive enzymatic hydrolysis steps and purified by the phenol-TEA-DOC extraction protocol. Ultrapure LPS-EK only activates the TLR4 pathway.