Learn More
Takara Bio Advantage® UltraPure PCR Deoxynucleotide Mix (10 mM each dNTP), 4 x 100 uL
SDP
Catalog No. NC9287432
Encompass
Advantage UltraPure PCR Nucleotide Mix, which contains all four dNTPs in a premixed aqueous solution, is ready for use in PCR amplification, cDNA synthesis, and in vitro mutagenesis. 4 x 100 uL
Catalog No. NC9287432 Supplier Takara Bio Supplier No. 639125
Advantage UltraPure PCR Nucleotide Mix, which contains all four dNTPs in a premixed aqueous solution, is ready for use in PCR amplification, cDNA synthesis, and in vitro mutagenesis. 4 x 100 uL