Learn More
Alpco Ltd Ultrasensitive Estradiol ELISA
SDP
Catalog No. NC2747274
Encompass_Preferred
The Ultrasensitive Estradiol ELISA¿is an enzyme immunoassay for the quantitative measurement of estradiol in serum or plasma (EDTA, heparin, or citrate plasma).¿ For¿In Vitro¿Diagnostic Use.
Catalog No. NC2747274 Supplier Alpco Ltd Supplier No. 20EDLHUUE01
The Ultrasensitive Estradiol ELISA¿is an enzyme immunoassay for the quantitative measurement of estradiol in serum or plasma (EDTA, heparin, or citrate plasma).¿ For¿In Vitro¿Diagnostic Use.