Learn More
Med Vet International ULTRASITE Non-Vented Two-Way Valve Bottle Top Attachment, J0463XA, Each
Catalog No. 5000581505
Encompass_Preferred
ULTRASITE Valve (Two-way) for aspiration or injection. DEHP-free. Latex-free. For attachment to standard 17mm or 20mm rubber-stoppered vials; non-vented.
Catalog No. 50-005-81505 Supplier Med Vet International Supplier No. JOR463XA
ULTRASITE Valve (Two-way) for aspiration or injection. DEHP-free. Latex-free. For attachment to standard 17mm or 20mm rubber-stoppered vials; non-vented.