Learn More
Electron Microscopy Sciences Ultra Smooth Carbon Adhesive Tabs, 12 mm
SDP
Catalog No. 5031334
Encompass_Preferred
Thin film of strong adhesive 1⁄2" diameter, over 99% transparent to EDS. With nickel (0.6%) and copper (< 0.3%). Used for gunshots residues. Small nonconductive particles can imaged and X-Rayed without conductiv coating.
Diameter: 12 mm
Catalog No. 50-313-34 Supplier Electron Microscopy Sciences Supplier No. 7782712
Thin film of strong adhesive 1⁄2" diameter, over 99% transparent to EDS. With nickel (0.6%) and copper (< 0.3%). Used for gunshots residues. Small nonconductive particles can imaged and X-Rayed without conductiv coating.
Diameter: 12 mm