Learn More
Alkali Scientific UltraSOFT 1000™ Nitrile Gloves, Fingertip Textured, Powder Free, 1000/Case, Extra Small
SDP
Catalog No. 5000662963
Encompass_Preferred
UltraSOFT 1000™ Nitrile Gloves provide outstanding comfort, durability, and barrier protection for professionals working in healthcare, laboratories, food processing, and industrial environments. Designed for extended wear, these powder free, non sterile gloves combine high tensile strength with excellent elasticity to maintain dexterity and consistent performance during demanding tasks. The accelerator free formulation helps reduce the risk of both Type I (latex protein) and Type IV (chemical accelerator) allergies, making them suitable for users who wear gloves for long periods. Textured fingertips improve grip and control in both wet and dry conditions. Tested to ASTM D6978 standards for chemotherapy drugs and fentanyl/opioid exposure, these gloves offer reliable protection when handling hazardous substances in clinical, pharmaceutical, and emergency response settings while meeting strict international quality and safety standards.
Catalog No. 50-006-62963 Supplier Alkali Scientific Supplier No. NG1105
UltraSOFT 1000™ Nitrile Gloves provide outstanding comfort, durability, and barrier protection for professionals working in healthcare, laboratories, food processing, and industrial environments. Designed for extended wear, these powder free, non sterile gloves combine high tensile strength with excellent elasticity to maintain dexterity and consistent performance during demanding tasks. The accelerator free formulation helps reduce the risk of both Type I (latex protein) and Type IV (chemical accelerator) allergies, making them suitable for users who wear gloves for long periods. Textured fingertips improve grip and control in both wet and dry conditions. Tested to ASTM D6978 standards for chemotherapy drugs and fentanyl/opioid exposure, these gloves offer reliable protection when handling hazardous substances in clinical, pharmaceutical, and emergency response settings while meeting strict international quality and safety standards.