Learn More
National Safety Apparel™ UltraSoft FR Balaclava
SDP
Catalog No. 17001742 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Color: Navy
Catalog No.
17-001-742 Color: Navy
1 options
Catalog No. 17-001-742 Supplier National Safety Apparel™ Supplier No. H73RY
Designed for comfort and flexibility, specifically for use with Arc Goggles. Inherently flame resistant.
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 6.5 oz. Westex™ UltraSoft™ Rib
  • Designed for use with Arc Goggles
  • Flat lock seam for comfort when worn under a hard hat
  • Designed for comfort and flexibility
  • FR fabric will not melt, drip, or ignite
  • Inherently flame resistant, FR protection will not wash/wear out
  • Lightweight, breathable and durable fabric
  • Made in the USA
Specifications
Size Medium
Material Cotton, Nylon
Color Navy
Seam Type Stitched
Certifications/Compliance CAT 2, NFPA 70 E, NFPA 2112, ASTM F1506, CSA Z462, OSHA 1910.269