Learn More
CUBE BIOTECH Ultrasolute™ Amphipol 17; 1 g
Catalog No. NC2684569
Encompass
1g of nanodisc copolymer Ultrasolute Amphipol 17 for the stabilization of membrane proteins outside the cell
Catalog No. NC2684569 Supplier CUBE BIOTECH Supplier No. 18302
Add to Cart
Edge
Add to Cart
1g of nanodisc copolymer Ultrasolute Amphipol 17 for the stabilization of membrane proteins outside the cell