Learn More
CUBE BIOTECH Ultrasolute™ Amphipol 18; 1 g
Catalog No. NC2684572
Encompass
1g of nanodisc copolymer Ultrasolute Amphipol 18 for the stabilization of membrane proteins outside the cell
Catalog No. NC2684572 Supplier CUBE BIOTECH Supplier No. 18322
1g of nanodisc copolymer Ultrasolute Amphipol 18 for the stabilization of membrane proteins outside the cell