Learn More
BRANSON ULTRASONICS CORPORATION (3P) ULTRASON SOL CONC GP 1QT 12/CS
Catalog No. NC3721572
Encompass_Preferred
Branson Ultrasonics Ultrasonic General-Purpose Cleaning Solutions, Size: 1 qt., Concentration: 10 to 12%, For Use With: Soils, fingerprints, tissue, light Oils, pH Range: 11 to 12
Catalog No. NC3721572 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 000955014
Branson Ultrasonics Ultrasonic General-Purpose Cleaning Solutions, Size: 1 qt., Concentration: 10 to 12%, For Use With: Soils, fingerprints, tissue, light Oils, pH Range: 11 to 12