Learn More
Andwin Scientific ULTRASONIC BATH 1800 DTH EA
SDP
Catalog No. 50727442
Encompass_Preferred
The Bransonic 1800 120V Digital Ultrasonic Bath features precision cleaning, power tracking technology that adjusts to load type, temperature customization, auto shut-off, sleep mode and solution degassing or conditioning; sold per piece. Tank size (L,W,D): 6 in.x5.5 in.x4 in. 150x140x100mm; Overall size (L,W,D): 9.9 in.x12 in.x11.9 in. 251x304x302mm; Weight: 7 lbs (3.2 kg); Frequency: 40kHz; Tank Capacity: .5 gal (1.9L); Drain: No
Catalog No. 50-727-442 Supplier Andwin Scientific Supplier No. 71020DTH
The Bransonic 1800 120V Digital Ultrasonic Bath features precision cleaning, power tracking technology that adjusts to load type, temperature customization, auto shut-off, sleep mode and solution degassing or conditioning; sold per piece. Tank size (L,W,D): 6 in.x5.5 in.x4 in. 150x140x100mm; Overall size (L,W,D): 9.9 in.x12 in.x11.9 in. 251x304x302mm; Weight: 7 lbs (3.2 kg); Frequency: 40kHz; Tank Capacity: .5 gal (1.9L); Drain: No